ADVH7F37FC48 AChemBlock

2-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carboxylic acid

  • MDL number MFCD11976234
  • Cas Number : 1260777-52-3
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-imidazo-1-2-a-pyridine-7-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32031
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart