ADVH7F37FC4B AChemBlock

8-(trifluoromethyl)isoquinolin-3-amine

  • MDL number MFCD22380387
  • Cas Number : 1357945-83-5
  • Molecular Weight : 212.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-trifluoromethyl-isoquinolin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32034
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart