ADVH7F37FC55 AChemBlock

5-chloro-8-fluoropyrido[3,4-b]pyrazine 5-chloro-8-fluoropyrido[4,3-b]pyrazine

  • MDL number MFCD22380480
  • Cas Number : 1374652-17-1
  • Molecular Weight : 183.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-8-fluoropyrido-3-4-b-pyrazine-5-chloro-8-fluoropyrido-4-3-b-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32048
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart