ADVH7F37FC56 AChemBlock

2-(1-benzyl-1H-pyrazol-3-yl)acetic acid

  • MDL number MFCD24485786
  • Cas Number : 1194803-69-4
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-1-benzyl-1h-pyrazol-3-yl-acetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32049
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart