ADVH7F37FC59 AChemBlock

2-chloro-4`-(trifluoromethyl)biphenyl-3-amine

  • MDL number MFCD18422904
  • Cas Number : 1261747-48-1
  • Molecular Weight : 271.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-trifluoromethyl-biphenyl-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32055
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart