ADVH7F37FC5B AChemBlock

(E)-2-aminobenzaldehyde O-methyl oxime

  • MDL number MFCD18824347
  • Cas Number : 305811-22-7
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/e-2-aminobenzaldehyde-o-methyl-oxime.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32057
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart