ADVH7F37FC5F AChemBlock

4-((4-iodophenoxy)methyl)benzamide

  • MDL number MFCD10367684
  • Cas Number : 1011018-52-2
  • Molecular Weight : 353.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-4-iodophenoxy-methyl-benzamide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32061
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart