ADVH7F37FC6F AChemBlock

7-bromo-2-methylisoquinolin-1(2H)-one

  • MDL number MFCD22380537
  • Cas Number : 1290634-35-3
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-methylisoquinolin-1-2h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32080
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart