ADVH7F37FC70 AChemBlock

1-fluoro-3-(difluoromethyl)-2-iodobenzene

  • MDL number MFCD18396819
  • Cas Number : 1261647-51-1
  • Molecular Weight : 272.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-fluoro-3-difluoromethyl-2-iodobenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32081
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart