ADVH7F37FC74 AChemBlock

1-(3-fluoro-2-iodophenyl)propan-1-one

  • MDL number MFCD18397044
  • Cas Number : 1261828-13-0
  • Molecular Weight : 278.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-fluoro-2-iodophenyl-propan-1-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32085
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart