ADVH7F37FC78 AChemBlock

6-bromo-2-cyclopropyl-5-methylbenzo[d]oxazole

  • MDL number MFCD19377860
  • Cas Number : 1267224-21-4
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-cyclopropyl-5-methylbenzo-d-oxazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32089
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart