ADVH7F37FC8B AChemBlock

furo[3,2-c]pyridine-2-carbaldehyde

  • MDL number MFCD15143903
  • Cas Number : 112372-07-3
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/furo-3-2-c-pyridine-2-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32117
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart