ADVH7F37FC90 AChemBlock

2-(1-methylpiperidin-4-yloxy)-5-(trifluoromethyl)benzenamine

  • MDL number MFCD03069457
  • Cas Number : 494777-68-3
  • Molecular Weight : 274.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-1-methylpiperidin-4-yloxy-5-trifluoromethyl-benzenamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32123
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart