ADVH7F37FC97 AChemBlock

2-(2-methoxyethyl)phenylboronic acid

  • MDL number MFCD22380339
  • Cas Number : 1122568-03-9
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-methoxyethyl-phenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32131
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart