ADVH7F37FC9F AChemBlock

1-ethynyl-2-(methylsulfonyl)benzene

  • MDL number MFCD20037596
  • Cas Number : 1019928-25-6
  • Molecular Weight : 180.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-ethynyl-2-methylsulfonyl-benzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32142
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart