ADVH7F37FCB3 AChemBlock

N,N-dimethyl(1-methylpiperidin-4-yl)methanamine dihydrochloride

  • MDL number MFCD09998358
  • Cas Number : 1170171-39-7
  • Molecular Weight : 229.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethyl-1-methylpiperidin-4-yl-methanamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32167
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart