ADVH7F37FCB4 AChemBlock

4-(benzofuran-2-yl)piperidine hydrochloride

  • MDL number MFCD05861403
  • Cas Number : 54402-12-9
  • Molecular Weight : 237.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-benzofuran-2-yl-piperidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32168
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart