ADVH7F37FCB5 AChemBlock

2-(2-ethylcyclopropyl)acetic acid

  • MDL number MFCD19229743
  • Cas Number : 344329-88-0
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-ethylcyclopropyl-acetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32172
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart