ADVH7F37FCBA AChemBlock

3-fluoro-5-hydroxypyridine-2-carbonitrile

  • MDL number MFCD18261780
  • Cas Number : 1211584-91-6
  • Molecular Weight : 138.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-5-hydroxypyridine-2-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32177
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart