ADVH7F37FCBC AChemBlock

1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-c]pyridin-3-amine

  • MDL number MFCD22689811
  • Cas Number : 1416712-70-3
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-tetrahydro-2h-pyran-2-yl-1h-pyrazolo-3-4-c-pyridin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32181
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart