ADVH7F37FCBF AChemBlock

1-ethyl-3-hydroxy-2-methylpyridin-4(1H)-one

  • MDL number MFCD01690785
  • Cas Number : 30652-12-1
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-ethyl-3-hydroxy-2-methylpyridin-4-1h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32184
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart