ADVH7F37FCC0 AChemBlock

N-((5-bromopyridin-3-yl)methyl)ethanamine dihydrochloride

  • MDL number MFCD22380372
  • Cas Number : 1337882-65-1
  • Molecular Weight : 288.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-5-bromopyridin-3-yl-methyl-ethanamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32185
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart