ADVH7F37FCC7 AChemBlock

ethyl 4-chloro-2,2-dimethylbutanoate

  • MDL number MFCD20622077
  • Cas Number : 53840-29-2
  • Molecular Weight : 178.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-chloro-2-2-dimethylbutanoate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32194
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart