ADVH7F37FCCC AChemBlock

N1-((3,5-dimethyl-1H-pyrazol-4-yl)methyl)ethane-1,2-diamine trihydrochloride

  • MDL number MFCD22380377
  • Cas Number : 1337879-50-1
  • Molecular Weight : 277.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n1-3-5-dimethyl-1h-pyrazol-4-yl-methyl-ethane-1-2-diamine-trihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32199
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart