ADVH7F37FCD5 AChemBlock

3-ethyl-5-(thiophen-3-yl)-1H-pyrazol-4-amine

  • MDL number MFCD22380360
  • Cas Number : 1257877-25-0
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-ethyl-5-thiophen-3-yl-1h-pyrazol-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32210
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart