ADVH7F38045E AChemBlock

1-(2-Methylphenyl)-cyclopropanamine

  • MDL number MFCD07374471
  • Cas Number : 503417-29-6
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/1-2-methylphenyl-cyclopropanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P37816
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart