ADVH7F380893 AChemBlock

2-(1-Pyrrolidinyl)-4-pyridinamine

  • MDL number MFCD13190680
  • Cas Number : 35981-63-6
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-1-pyrrolidinyl-4-pyridinamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P38560
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart