ADVH7F380B47 AChemBlock

1-(4-Piperidinylmethyl)-2-pyrrolidinone hydrochloride

  • MDL number MFCD20731152
  • Cas Number : 139726-90-2
  • Molecular Weight : 218.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/1-4-piperidinylmethyl-2-pyrrolidinone-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41391
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart