ADVH7F38110F AChemBlock

N-Boc-N-cyclohexyl-glycine

  • MDL number MFCD16620245
  • Cas Number : 166739-13-5
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/n-boc-n-cyclohexyl-glycine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P40718
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart