ADVH7F3812B1 AChemBlock

11-(Boc-amino)-1-undecyl bromide

  • MDL number MFCD22576479
  • Cas Number : 463930-53-2
  • Molecular Weight : 350.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/11-boc-amino-1-undecyl-bromide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P37979
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart