ADVH7F381384 AChemBlock

methyl 3-fluoro-6-methylpyridine-2-carboxylate

  • MDL number MFCD18258174
  • Cas Number : 1256822-15-7
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-fluoro-6-methylpyridine-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43479
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart