ADVH7F3813CB AChemBlock

3-(aminomethyl)-1-benzylpyridin-2(1H)-one

  • MDL number MFCD26696351
  • Cas Number : 933757-00-7
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-1-benzylpyridin-2-1h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43173
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart