ADVH7F3813FD AChemBlock

methyl 4,5-dichloropyridine-3-carboxylate

  • MDL number MFCD16038783
  • Cas Number : 343781-51-1
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-5-dichloropyridine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43294
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart