ADVH7F38141D AChemBlock

(E)-N`-(2,4-dimethylphenyl)-N-methylformimidamide hydrochloride

  • MDL number MFCD00463812
  • Cas Number : 51550-40-4
  • Molecular Weight : 198.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/e-n-2-4-dimethylphenyl-n-methylformimidamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43258
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart