ADVH7F38141E AChemBlock

(1S,3R)-dimethyl cyclopentane-1,3-dicarboxylate

  • MDL number MFCD00275730
  • Cas Number : 190137-08-7
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1s-3r-dimethyl-cyclopentane-1-3-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43347
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart