ADVH7F381485 AChemBlock

tert-butyl trans-2,2-dimethyl-3-hydroxycyclobutylcarbamate

  • MDL number MFCD27956963
  • Cas Number : 1434141-89-5
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/tert-butyl-trans-2-2-dimethyl-3-hydroxycyclobutylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P48173
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart