ADVH7F38150B AChemBlock

2-methylsulfanylpyrido[4,3-d]pyrimidine

  • MDL number MFCD28662034
  • Cas Number : 1554485-62-9
  • Molecular Weight : 177.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methylsulfanylpyrido-4-3-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47873
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart