ADVH7F3815A5 AChemBlock

tert-butyl N-[exo-2-azabicyclo[2.2.1]heptan-5-yl]carbamate

  • MDL number MFCD31707754
  • Cas Number : 1932203-04-7
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-exo-2-azabicyclo-2-2-1-heptan-5-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46653
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart