ADVH7F38172F AChemBlock

4-chloro-7-fluoro-1H-pyrazolo[4,3-c]pyridine

  • MDL number MFCD22380482
  • Cas Number : 1374651-76-9
  • Molecular Weight : 171.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/4-chloro-7-fluoro-1h-pyrazolo-4-3-c-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48558
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart