ADVH7F3818DE AChemBlock

1H-Pyrazole-3,5-dicarbonitrile

  • MDL number MFCD28168072
  • Cas Number : 847573-68-6
  • Molecular Weight : 118.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/1h-pyrazole-3-5-dicarbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=P51498
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart