ADVH7F381923 AChemBlock

endo-2,3-NorbornanedicarboxiMide

  • MDL number MFCD13193124
  • Cas Number : 28871-95-6
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/endo-2-3-norbornanedicarboximide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P51588
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart