ADVH7F381C99 AChemBlock

2-(1,1-difluoroethyl)-5-methyl-N-(4-(pentafluoro-l6-sulfaneyl)phenyl)-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD28502125
  • Cas Number : 1282041-94-4
  • Molecular Weight : 415.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-1-1-difluoroethyl-5-methyl-n-4-pentafluoro-l6-sulfaneyl-phenyl-1-2-4-triazolo-1-5-a-pyrimidin-7-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65520
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart