ADVH7F38293D AChemBlock

3-(4-methoxyphenyl)cyclopentan-1-one

  • MDL number MFCD06659800
  • Cas Number : 116526-34-2
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/3-4-methoxyphenyl-cyclopentan-1-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=Q69545
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart