ADVH7F382A62 AChemBlock

6-chloro-4-nitro-1H-indole

  • MDL number MFCD07781454
  • Cas Number : 245524-95-2
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/6-chloro-4-nitro-1h-indole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=Q69885
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart