ADVH7F3831AD AChemBlock

(4-(ethoxymethyl)-3-fluorophenyl)boronic acid

  • MDL number MFCD14687264
  • Cas Number : 1334218-48-2
  • Molecular Weight : 198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/4-ethoxymethyl-3-fluorophenyl-boronic-acid.html

Safety Information

signalword Warning
pcodes P261; P264; P270; P271; P280; P301+P312; P302+P352; P304+P340; P330; P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86433
pictograms GHS07
hstatements H302; H312; H332
Availability details for
Quantity
  • Availability
  • Add to Cart