ADVH7F3831D8 AChemBlock

3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]pyridine

  • MDL number MFCD21099734
  • Cas Number : 1286202-05-8
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/3-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-pyridine.html

Safety Information

signalword Warning
pcodes P261; P305 + P351 + P338; P302 + P352
MSDS https://www.achemblock.com/ac_sds?Catalog=S81360
pictograms GHS07
hstatements H315; H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart