ADVH7F383267 AChemBlock

2,6-Dichloro-3H-pyrimidin-4-one

  • MDL number MFCD00955562
  • Cas Number : 120977-94-8
  • Molecular Weight : 164.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-6-dichloro-3h-pyrimidin-4-one.html

Safety Information

signalword Warning
pcodes P261; P305+P351+P338
MSDS https://www.achemblock.com/ac_sds?Catalog=S86439
pictograms GHS07
hstatements H315; H319; H320
Availability details for
Quantity
  • Availability
  • Add to Cart