ADVH7F3833C8 AChemBlock

2-(tributylstannyl)acetonitrile

  • MDL number MFCD32173435
  • Cas Number : 17729-59-8
  • Molecular Weight : 330.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at 0-8 �C
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/2-tributylstannyl-acetonitrile.html

Safety Information

signalword Danger
pcodes P210; P233; P240; P241; P242; P243; P260; P264; P270; P273; P280; P301+P310+P330; P303+PP361+P353; P305+P351+P338; P314; P362; P363; P370+P378
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72336
pictograms GHS02;GHS06;GHS08;GHS09
hstatements H226; H301; H312; H315; H319; H372
Availability details for
Quantity
  • Availability
  • Add to Cart