ADVH7F38340A AChemBlock

tert-butyl 3-hydroxymorpholine-4-carboxylate

  • MDL number MFCD32173463
  • Cas Number : 1097871-14-1
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/tert-butyl-3-hydroxymorpholine-4-carboxylate.html

Safety Information

signalword Warning
pcodes P301+312
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86928
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart