ADVH7F38366E AChemBlock

(S)-2-amino-2-cyclopentylethan-1-ol hydrochloride

  • MDL number MFCD30530442
  • Cas Number : 2055848-97-8
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/s-2-amino-2-cyclopentylethan-1-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261; P305 + P351 + P338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87765
pictograms GHS07
hstatements H302; H315; H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart